Learn More
anti-CD45.2, Alexa Fluor 488, Clone: 104, R&D Systems™
Manufacturer:  R&D Systems FAB8448G025
Catalog No. FAB8448G025
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
CD45.2
104
Flow Cytometry 5 uL/10^6 cells
Purified
PTPRC
B10.S mouse thymocytes and splenocytes
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
19264
Mouse
Flow Cytometry
Alexa Fluor 488
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
B220, CD 45, CD45, cd45 antigen, CD45.2, CD45R, GP 180, GP180, Human homolog of severe combined immunodeficiency due to PTPRC deficiency, L CA, LCA, Leukocyte common antigen, LY 5, Ly5, Ly5.2, Ly-5.2, Protein tyrosine phosphatase receptor type C, Protein tyrosine phosphatase receptor type c polypeptide, PTPRC, RT7.2, SCID due to PTPRC deficiency, T200, T200 glycoprotein, T200 leukocyte common antigen
Mouse
IgG2a
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects mouse cells expressing the CD45.2 allotype in flow cytometry. Clone 104 does not detect the mouse CD45.1 alloantigen.