Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD45.2 Red Fluor 710, Clone: 104,

Mouse Monoclonal Antibody

$153.50 - $272.50

Specifications

Antigen CD45.2
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF1475MI
View Documents
MilliporeSigma™
MABF1475
25μg
Each for $153.50
Add to cart
 
MABF1476MI
View Documents
MilliporeSigma™
MABF1476
100μg
Each for $272.50
Add to cart
 
Description

Description

Specifically detects CD45.2 Clone: 104 in Mouse samples, and it is validated for Flow Cytometry

The 104 antibody reacts with mouse CD45.2, also known as Ly5.2, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors. The 104 antibody is typically used as a leukocyte marker in Ly5.2 mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. The antibody has been demonstrated to be specific for CD45.2 and is not cross-reactive with CD45.1-bearing cells.
Specifications

Specifications

CD45.2
Monoclonal
Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Flow Cytometry
Proprietary immunogen
Ptprc, Ly-5
RUO
Purified by size-exclusion chromatography
Primary
Purified
104
Mouse
P06800
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Inflammation, Immunology
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit