Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Mouse, Unlabeled, Clone: 69, BD

Mouse Monoclonal Antibody

$213.00 - $458.00

Specifications

Antigen CD45
Isotype IgG1
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610265
View Documents
BD Biosciences
610265
50μg
Each for $213.00
Add to cart
 
BDB610266
View Documents
BD Biosciences
610266
150μg
Each for $458.00
Add to cart
 
Description

Description

CD45 (also known as leukocyte common antigen, GP180, or T200) is found on all hematopoietic cells except those of red cell origin. The N-terminal domain is a large glycosylated extracellular region of variable length (390-542 amino acids) derived by the alternative splicing of at least three exons (4, 5, or 6). This variation accounts for the different isoforms of CD45 (180-220 kDa). The remainder of the molecule includes a short transmembrane region followed by a large highly conserved cytoplasmic domain of 702 residues. The CD45 cytoplasmic domain possesses an intrinsic protein tyrosine phosphatase (PTPase) activity. This region contains two PTPase domains (domains I and II) which share 40% homology. Both domains are required for optimal PTPase activity and show significant homology with other receptor-like and cytoplasmic PTPases. CD45 plays a critical role in antigen receptor-induced responses of T and B cells. In B cells, CD45 can exist as part of a larger multisubunit complex composed of several tyrosine phosphorylated proteins, including the Src-family PTK, Lyn, or it can exist as the membrane immunoglobulin-associated MB-1/B29 heterodimer. In T cells, CD45 associates with and dephosphorylates both lck and fyn. In addition, CD45 can associate with membrane proteins involved in T cell activation, such as CD2, Thy-1, TCR, and CD26.

Specifications

Specifications

CD45
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Unconjugated
Immunology
Mouse
Affinity Purified
Monoclonal
Leukocyte Common Antigen; GP180; T200
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
69
250μg/mL
Store undiluted at -20°C.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit