Learn More
CD45, clone: BHPT-1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112129
Catalog No. 01-670-848
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD45 Leukocyte Common Antigen (LCA) belongs to the family of at least four isoforms of membrane glycoproteins (220, 205, 190, 180 kDa) expressed on hematopoietic cell lines but absent on non-hematopoietic cell lines, normal and malignant, non-hematopoietic tissues. The intracellular portion of these molecules have protein phosphatase activity and are involved in regulation of transmembrane signals.
Specifications
Specifications
CD45
Monoclonal
Unconjugated
PTPRC
Liquid
PTPRC
human peripheral blood monocytes from a rheumatoid arthritis patient
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
BHPT-1
PBS with 0.08% sodium azide
P08575
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Murine
IgG1
100μg
-20° C, Avoid Freeze/Thaw Cycles, Avoid Freeze/Thaw Cycles
Primary
5788