Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Mouse, Brilliant Violet 510, Clone: D058-1283, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 563530

Catalog No. BDB563530


Add to cart

Description

Description

D058-1283 is a CD45 monoclonal antibody specific for non-human primate leucocytes, developed using Rhesus peripheral whole blood as immunogen. It does not cross react with human leucocytes. This antibody reacts with Baboon, Rhesus and Cynomolgus Macaque leucocytes in a similar pattern as seen with CD45 binding to the Leukocyte Common Antigen on human cells. Immunophenotypic analysis shows that D058-1283 binds to lymphocytes, monocytes and granulocytes of non-human primate blood samples. This antibody is able to block the binding of monoclonal antibody TÜ116; a reported anti-human CD45 antibody that cross-reacts with nonhuman primate leucocytes. In Western blot analysis, the D058-1283 antibody identifies a band of approximately 180-200 kDa molecular weight .

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon™ BV510 can be excited by the violet laser and detected in the BD Horizon™ V500 (525/50-nm) filter set. BD Horizon™ BV510 conjugates are useful for the detection of dim markers off the violet laser.

Specifications

Specifications

CD45
D058-1283
Pan Leukocyte, NHP-specific
Affinity Purified
Rhesus peripheral whole blood
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Violet 510
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Baboon, Cynomolgus, Rhesus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit