Learn More
anti-CD45, PE, Clone: 2D1, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB1430P025
 View more versions of this product
Catalog No. FAB1430P025
Add to cart
Specifications
Specifications
CD45
2D1
Flow Cytometry 10 uL/10^6 cells
Supernatant
PTPRC
Human peripheral blood mononuclear cells
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5788
Human
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Mouse
IgG1
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human CD45. This antibody recognizes all isoforms of human CD45.