Learn More
anti-CD45, PE, Clone: 2D1, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB1430P100
 View more versions of this product
Catalog No. FAB1430P100
Add to cart
Description
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
CD45
2D1
Flow Cytometry 10 uL/10^6 cells
Supernatant
PTPRC
Human peripheral blood mononuclear cells
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5788
Human
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Mouse
IgG1
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human CD45. This antibody recognizes all isoforms of human CD45.