Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD45, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$471.50

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:KLENLEPEHEYKCDSEILYNNHKFTNASKIIKTDFGSPGEPQIIFCRSEAAHQGVITWNPPQRSFHNFTLCYIKETEKDCLNLDKNLIKYDLQNLKPYTKYVLSLHAYIIAKVQRNGSAAMCHFTTKSAPPSQVWNMT
Antigen CD45
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP188103
View Documents
Novus Biologicals
NBP188103
0.1 ml
Each for $471.50
Add to cart
 
Description

Description

CD45 Polyclonal antibody specifically detects CD45 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rabbit
This antibody was developed against Recombinant Protein corresponding to amino acids:KLENLEPEHEYKCDSEILYNNHKFTNASKIIKTDFGSPGEPQIIFCRSEAAHQGVITWNPPQRSFHNFTLCYIKETEKDCLNLDKNLIKYDLQNLKPYTKYVLSLHAYIIAKVQRNGSAAMCHFTTKSAPPSQVWNMT
Polyclonal
Primary
Immunohistochemistry, Immunohistochemistry (Paraffin)
5788
IgG
CD45
Affinity Purified
RUO
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit