Learn More
CD45 Mouse anti-Human, V450, Clone: 2D1, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 642275
Catalog No. BDB642275
Add to cart
Description
Description
CD45 clone 2D1 is derived from the hybridization of mouse NS-1 myeloma cells with spleen cells from BALB/c mice immunized with human peripheral blood mononuclear cells (PBMCs).
The CD45 antibody (anti–HLe-1) recognizes members of the T200 family of human leucocyte antigens with molecular mass of 180 to 220 kilodaltons (kDa).
Specifications
Specifications
CD45
2D1
V450
Conjugated
IgG1, κ
RUO
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Monoclonal
Flow Cytometry
100μg/mL
Phosphate buffered saline with gelatin and 0.1% sodium azide.
Mouse
100 Tests
Immunology
Primary
Human