Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RA Rat anti-Mouse, PerCP-Cy5.5, Clone: 14.8, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 564359

Catalog No. BDB564359


Add to cart

Description

Description

The 14.8 clone has been reported to react with an exon A-dependent epitope of the CD45 protein, which is found at high density on B cells and at low density on peripheral T cytotoxic/suppressor cells and a very small subset of thymocytes. Nearly all B-lineage cells, including B-cell precursors in fetal liver and adult bone marrow and Ig-secreting cells, but not hematopoietic stem cells or myeloid progenitors, have been reported to be detectable by mAb 14.8. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus, differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. mAb 14.8 has been reported to enhance the proliferative effect of PHA on purified spleen T cells, possibly by replacing a signal normally delivered by accessory cells, to enhance isotype switching during in vitro B-cell responses, and to inhibit antigen-induced p21 [ras] activation.

Specifications

Specifications

CD45RA
14.8
PerCP-Cyanine5.5
Aqueous buffered solution containing ≤0.09% sodium azide
Rat
IgG2b, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Ptprc; CD45R; CD45; LCA; Leukocyte common antigen; Ly-5; Lyt-4
Affinity Purified
Radiation-induced NZC mouse B lymphoma WEHI-279
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit