Learn More
CD45RA Mouse anti-Human, PE, Clone: BHKG-2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112135
Catalog No. 01-670-854
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing and these isoforms differ in their extracellular domains, however, they share identical transmembrane and cytoplasmic domains. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. Besides the role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45 is expressed on cells of the human hematopoietic lineage, with the exception of mature red cells and is not detected on differentiated cells of other tissues. It is likely that CD45 plays an important role in signal transduction, inhibition or upregulation of various immunological functions. Diseases associated with CD45 dysfunction include severe combined immunodeficiency and Hepatitis C.
Specifications
Specifications
CD45RA
Monoclonal
PE
PTPRC
Liquid
PTPRC
human peripheral blood monocytes from a rheumatoid arthritis patient
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
BHKG-2
PBS with 0.2% carrier protein and 0.08% sodium azide
P08575
B220, CD45, CD45R, GP180, LCA, LY5, T200
Mouse
IgG1
100 Tests
4° C, do not freeze
Primary
5788