Learn More
CD45RO Mouse anti-Human, Alexa Fluor 700, Clone: UCHL1, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 561136
Catalog No. BDB561136
Add to cart
Description
Description
The UCHL1 monoclonal antibody specifically binds to the 180 kDa isoform of CD45 (aka, the Leukocyte Common Antigen). CD45RO is a type I transmembrane glycoprotein that has cytoplasmic protein tyrosine phosphatase activity and functions in signal transduction pathways. This CD45 isoform does not include amino acid sequences encoded by the variable CD45 exons A, B, or C. CD45RO is expressed on most thymocytes, activated T cells, memory T cells, granulocytes and monocytes, but only on a proportion of resting T cells. CD45RO and CD45RA antibodies seem to define complementary, predominantly non-overlapping, populations in resting peripheral T cells, demonstrating heterogeneity within the CD8 and CD4 subpopulations. CD45RO binds to CD22.
Specifications
Specifications
CD45RO
UCHL1
CD45R; PTPRC; LCA; Leukocyte common antigen; GP180; LY5; T200
Affinity Purified
Human IL-2-dependent T-cell line
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Alexa Fluor 700
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Mouse
IgG2a, κ
RUO
Primary
Human