Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD47, Mouse anti-Human, Clone: B6H12, BV786, BD

Manufacturer:  BD Biosciences 563758

Catalog No. BDB563758


Add to cart

Description

Description

Host Species: Mouse
Clone: B6H12
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Purified human placental RGD-binding proteins

Flow cytometry

Specifications

Specifications

CD47
Affinity Purified
Purified human placental RGD-binding proteins
RUO
Monoclonal
B6H12
Murine
100 Tests
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit