Learn More
anti-CD48/SLAMF2, PE, Clone: 394607, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Manufacturer:  R&D Systems FAB3644P
Catalog No. FAB3644P
Add to cart
This item is not returnable. View return policy
Description
Description
CD48/SLAMF2 Monoclonal specifically detects CD48/SLAMF2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD48/SLAMF2
394607
Flow Cytometry 10 uL/10^6 cells
P09326.2
BCM1, BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2
Mouse
IgG2b
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human CD48/SLAMF2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CD48 or recombinant human OX40 is observed.
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CD48
Mouse myeloma cell line NS0-derived recombinant human CD48/SLAMF2 Gln27-Ser220 Accession # P09326.2
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
962
Human