Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49b Mouse anti-Human, Alexa Fluor 647, Clone: AK-7, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564314

Catalog No. BDB564314


Add to cart

Description

Description

The AK-7 monoclonal antibody specifically binds to CD49b. CD49b is also known as integrin α2 chain, VLA-α2, and GP1a. CD49b is a type I transmembrane glycoprotein and be longs to the integrin family of extracellular matrix and cell-cell adhesion receptors. CD49b is expressed on numerous cell types including platelets, activated T cells, B cells, and monocytes. CD49b noncovalently associates with CD29 to form the VLA-2 complex (integrin α 2/ β 1). The VLA-2 complex serves as a receptor for collagen and laminin. It can regulate responses mediated by proinflammatory effector cells.

Specifications

Specifications

CD49b
AK-7
Integrin α2 chain; VLA-α2: GP1a
Affinity Purified
IgG1, κ
RUO
Primary
Human
Flow Cytometry
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Murine
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit