Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49d Rat anti-Mouse, Brilliant Violet 786, Clone: R1-2, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 564397

Catalog No. BDB564397


Add to cart

Description

Description

The R1-2 monoclonal antibody specifically binds to α 4 chain (CD49d), which is expressed as a heterdimer with either of two β , β 1 or β 7 (also known as β p). The α 4β α 4β 7 also preferentially binds to the mucosal vascular addressin, MAdCAM-1. The R1-2 antibody blocks some α 4 integrin-mediated binding functions. In combination with mAb 9C10 (MFR4.B) (Cat. No. 553313), binding of VLA-4 expressing cells to VCAM-1 can be almost completely inhibited.

Specifications

Specifications

CD49d
R1-2
Brilliant Violet 786
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
CDF IgG2b, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Integrin α4; Itga4; Integrin alpha-4; LPAM alpha; VLA-4 alpha; VLA-4a
Affinity Purified
AKR/Cum mouse spontaneous T lymphoma line TK1
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit