Learn More
CD49D Mouse anti-Human, Brilliant Violet 510, Clone: 9F10, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 563458
Catalog No. BDB563458
Add to cart
Description
Description
The 9F10 monoclonal antibody specifically reacts with the integrin α 4 chain, that is expressed as a heterodimer with either of two β integrin subunits, β 1 (CD29) or β 7. The α 4β 1 integrin (VLA-4) is expressed on lymphocytes, monocytes, thymocytes, NK cells, and several B- and T-cell lines, and mediates binding to VCAM-1 (CD106) and the CS-1 region of fibronectin. The α β 7 also binds fibronectin and VCAM-1, and has been shown in the mouse to preferentially bind the mucosal vascular addressin molecule, MAdCAM-1. This antibody is useful for studies of the expression by and function of cells that express α 4 chain-containing integrins. This clone cross-reacts with a subset of peripheral blood lymphocytes, monocytes, and some granulocytes of baboon and both rhesus and cynomolgus macaque monkeys. The distribution on leukocytes is similar to that observed with human peripheral blood leukocytes.
Specifications
Specifications
CD49d
9F10
Integrin α4 chain; Integrin alpha 4; ITGA4; IA4; alpha 4 subunit of VLA-4
Affinity Purified
IgG1, κ
RUO
Primary
Baboon, Bovine, Canine, Cynomolgus, Human, Rhesus
Flow Cytometry
Brilliant Violet 510
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal