Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49e Rat anti-Mouse, Alexa Fluor 647, Clone: 5H10-27 (MFR5), BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 564312

Catalog No. BDB564312


Add to cart

Description

Description

The 5H10-27 (MFR5) monoclonal antibody specifically binds to the α5 chain of the integrin α5β1 fibronectin receptor (CD49e/CD29, VLA-5) on thymocytes, activated T lymphocytes, precursor and activated B cells, mast cells, platelets, epithelial and endothelial cells, fibroblasts and a variety of mouse cell lines. Soluble 5H10-27 (MFR5) antibody has been reported to inhibit VLA-5-mediated functions in vitro. In addition, immobilized 5H10-27 (MFR5) antibody reportedly costimulates the proliferative response of CD8+ T cells to plate-bound anti-CD3e antibody.

Specifications

Specifications

CD49e
5H10-27 (MFR5)
Alexa Fluor 647
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Itga5; ITA5; Integrin alpha-5; Fibronectin receptor alpha; Fnra; VLA5 alpha
Affinity Purified
Mouse Mast Cell Line MC/9
100μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit