Learn More
CD49e Rat anti-Mouse, Biotin, Clone: 5H10.27, Southern Biotech™
SDP
Rat Monoclonal Antibody
Manufacturer:  Southern Biotechnology Associates 169508
Catalog No. OB169508
Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
Specifications
CD49e
Monoclonal
0.5mg/mL
Supplied in PBS/NaN3
C57BL/6 x A/J F1 mouse mast cell line MC/9
RUO
Mouse
IgG2a
Flow Cytometry, Immunohistochemistry (Frozen)
5H10.27
Biotin
Rat
0.5 mg
Primary
Store at 2-8°C