Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512507

Catalog No. PIPA512507


Add to cart

Description

Description

CD49e belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein is the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
Specifications

Specifications

ITGA5
Polyclonal
PBS with 0.09% sodium azide
P08648, P11688
FNRA, CD49e, VLA5A, ITGA5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
16402, 3678
Flow Cytometry, Western Blot
Unconjugated
ITGA5
Liquid
ITGA5
KLH conjugated synthetic peptide between 796-824 amino acids from the C-terminal region of human CD49e
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit