Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD49f; Clone: GoH3; BD

Manufacturer:  BD Biosciences 563271

Catalog No. BDB563271


Add to cart

Description

Description

Host Species: Rat
Clone GoH3
Isotype IgG2a, κ
Species Reactivity Hu
Immunogen Mouse mammary tumor cells

Flow Cytometry

Specifications

Specifications

CD49f
Brilliant Violet 510
Rat
IgG2a
RUO
Monoclonal
GoH3
Affinity Purified
Mouse mammary tumor cells
50 Tests
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit