Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49f Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526262

Catalog No. PIPA526262


Add to cart

Description

Description

This antibody is predicted to react with bovine, porcine and rat based on sequence homology.

The CD49f protein product is the integrin alpha chain alpha 6. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. For example, alpha 6 may combine with beta 4 in the integrin referred to as TSP180, or with beta 1 in the integrin VLA-6. Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling.
Specifications

Specifications

CD49f
Polyclonal
PBS with 0.09% sodium azide
P23229
CD49f, ITGA6B, VLA-6, CD49 antigen-like family member F, integrin alpha-6, ITGA6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3655
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
ITGA6
Liquid
ITGA6
KLH conjugated synthetic peptide between 1082-1110 amino acids from the C-terminal region of human CD49f
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit