Learn More
CD5 Mouse anti-Human, APC, Clone: M28623, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112255
Catalog No. 01-670-951
Add to cart
Description
Description
CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter’s Syndrome.
Specifications
Specifications
CD5
Monoclonal
APC
CD5
Liquid
CD5
human T-acute lymphoblastic leukemia (ALL) cells
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
M28623
PBS with 0.2% carrier protein and 0.08% sodium azide
P06127
CD5 antigen p56-62, T-cell surface glycoprotein CD5, lymphocyte antigen T1/Leu-1
Mouse
IgG2a
100 Tests
4° C, do not freeze
Primary
921