Learn More
anti-CD5, BB515; Clone: UCHT2; BD
Manufacturer:  BD Biosciences 564647
Catalog No. BDB564647
Add to cart
Description
Description
Applications:
Flow cytometry
Specifications
Specifications
CD5
UCHT2
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Murine
IgG1, kappa
Store undiluted at 4deg.C and protected from prolonged exposure to light. Do not freeze
Monoclonal
Flow Cytometry
BB515
Affinity purified
Human T cells
RUO
Primary
Human