Learn More
CD5, BUV737, Mouse anti-Human, Clone: UCHT2, BUV737, BD
Manufacturer:  BD Biosciences 564452
Catalog No. BDB564452
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Applications:
Flow cytometry
Specifications
Specifications
CD5
UCHT2
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Mouse
IgG1, kappa
RUO
Primary
Human
Flow Cytometry
Brilliant Ultraviolet 737
Affinity purified
Human T cells
25 Tests
Store undiluted at 4deg.C and protected from prolonged exposure to light. Do not freeze
Monoclonal