Learn More
CD5/CD19 Mouse anti-Human, FITC, PE, Invitrogen™
Antibody Cocktail
Manufacturer:  Invitrogen MA112136
Catalog No. 01-670-855
Add to cart
Description
Description
This antibody is a monoclonal mixture of FITC-conjugated CD5 (clone M28623, isotype IgG2a) and PE-conjugated CD19 (clone 1G9, isotype IgG1).
In humans, CD5 is a 67kDa T lymphocyte single chain transmembrane glycoprotein. It is present on all mature T lymphocytes, on most of thymocytes and on many T cell leukemias and lymphomas. It reacts with a subpopulation of activated B cells.
Specifications
Specifications
CD5/CD19
Cocktail
FITC, PE
CD19
Liquid
CD19, CD5
human T-acute lymphoblastic leukemia (ALL) cells
Protein A/G
RUO
Antibody
921, 930
Flow Cytometry
1G9
PBS with 0.2% carrier protein and 0.08% sodium azide
P06127, P15391
CD5 antigen p56-62, T-cell surface glycoprotein CD5, lymphocyte antigen T1/Leu-1, AW495831, B-lymphocyte antigen CD19, differentiation antigen CD19
Mouse
IgG1/IgG2a
100 Tests
4° C, do not freeze
Primary
Human