Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137292

Catalog No. 01-673-207

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CD5 is a 67kDa human T-lymphocyte single-chain transmembrane glycoprotein. It is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. It also reacts with a subpopulation of activated B-cells.

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is tonsil tissue.
Specifications

Specifications

CD5
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
P06127
CD5 antigen p56-62, T-cell surface glycoprotein CD5, lymphocyte antigen T1/Leu-1
Rabbit
IgG
1mL
4° C, do not freeze
Primary
921
Immunohistochemistry (Paraffin)
Unconjugated
CD5
Liquid
CD5
Synthetic peptide from the intracellular region of CD5
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit