Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD50 Mouse anti-Human, PE, Clone: TU41 (also known as TÜ41), BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564209

Catalog No. BDB564209


Add to cart

Description

Description

The TU41 monoclonal antibody specifically binds to the intracellular adhesion molecule-3 (ICAM-3) which is also known as CD50. ICAM-3 is a 110-120 kDa type I membrane glycoprotein expressed on all leukocytes (platelets and erythrocytes are ICAM-3-negative). ICAM-3 is also expressed on epidermal Langerhans cells and endothelial cells. It predominantly interacts with LFA-1 (CD11a/CD18) without requiring activation as the initial dendritic cell-T lymphocyte interaction. ICAM-3 is an important LFA-1 ligand that functions in the initiation of the immune/inflammatory response and also plays a role in chemotaxis.

Specifications

Specifications

CD50
TU41 (also known as TÜ41)
ICAM-3; ICAM3; Intercellular adhesion molecule 3
Affinity Purified
IgG2b, κ
RUO
Primary
Human
Flow Cytometry
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit