Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD51 Mouse, Unlabeled, Clone: 21, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611013

 View more versions of this product

Catalog No. BDB611013


Add to cart

Description

Description

Integrins are a family of proteins that mediate intercellular adhesion or adherence to extracellular matrix proteins. Their roles are essential for embryonic development, tumor metastasis, organ function, and proper immune cell function. Family members are heterodimers that contain a larger α subunit that is unique to each individual receptor and a smaller β subunit that can be shared by several receptors. Based on β subunit content, integrins are divided into the subfamilies β1, β2, and β3. The β3 subfamily contains the vitronectin receptor (αVβ3) and the platelet protein gpIIb/IIIa. The α chain of the vitronectin receptor (VNRα, CD51, integrin αV) has been reported to consist of a disulfide-linked large extracellular (125kDa) subunit and a smaller (25kDa) membrane-anchored subunit. The large subunit contains multiple sequences with homology to calcium-binding sites in other proteins and an RGD-dependent ligand-binding site, while the small subunit contains a transmembrane domain and a short cytoplasmic domain. VNRα has been reported to be noncovalently associated with the VNR β3-chain (CD61, gpIIIa). The VNR mediates cell adhesion to RGD-containing ligands such as vitronectin, von Willebrand factor, fibrinogen, and thrombospondin.

Immunofluorescence, Western Blotting

Specifications

Specifications

CD51
21
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Integrin αV; Vitronectin Receptor α; VNRα
Affinity Purified
Human CD51/VNRα aa. 609-722
150μg
Immunology
Primary
Canine, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.