Learn More
ICAM-1 Mouse anti-Human, Biotin, Clone: HM.1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA135240
Catalog No. 01671203
Add to cart
Description
Description
ICAM-1 (CD54) is an 85-110 kDa single-chain type 1 integral membrane glycoprotein with an extracellular domain of five immunoglobulin superfamily repeats, a transmembrane region and a cytoplasmic domain. ICAM-1 has 7 potential N-linked glycosylation sites and shares considerable amino acid sequence homology with ICAM-3 (CD50) and ICAM-2 (CD102). ICAM-1 binds to integrins of type CD11a/CD18 (leukocyte adhesion molecule, LFA-1), or CD11b/CD18 (Mac-1) and is exploited by Rhinovirus as a receptor. ICAM-1 is expressed by activated endothelial cells and detected on epithelial cells, fibroblasts, chondrocytes, B lymphocytes, T lymphocytes (low), monocytes, macrophages, dendritic cells and neutrophils, with lower levels that increase upon inflammation. ICAM-1 is also detected in some carcinoma and melanoma cells. Soluble ICAM-1 is detectable in the plasma and is elevated in patients with various inflammatory syndromes.
Specifications
Specifications
ICAM-1
Monoclonal
Biotin
ICAM1
Liquid
ICAM1
ICAM1.
Purified
RUO
Antibody
Monoclonal
Human
Immunofluorescence, Immunohistochemistry (Frozen)
HM.1
PBS with 0.1% BSA and 0.02% sodium azide
P05362
BB2, P3.58, ICAM-1, cell surface glycoprotein P3.58, intercellular adhesion molecule 1, human rhinovirus receptor, major group rhinovirus receptor
Mouse
IgG1
50 μg
4° C
Primary
3383