Learn More
ICAM-1 Mouse anti-Human, Clone: MEM-112, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA119125
Catalog No. 01-675-897
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD54 (ICAM-1) is a 90 kD member of the C2 subset of immunoglobulin superfamily. It is a transmembrane molecule with 7 potential N-glycosylated sites, expressed on resting monocytes and endothelial cells and can be upregulated on many other cells, e.g. with lymphokines, on B- and T-lymphocytes, thymocytes, dendritic cells and also on keratinocytes, chondrocytes, as well as epithelial cells. CD54 mediates cell adhesion by binding to integrins CD11a/CD18 (LFA-1) and to CD11b/CD18 (Mac-1). The interaction of CD54 with LFA-1 enhances antigen-specific T-cell activation.
Specifications
Specifications
ICAM-1
Monoclonal
Unconjugated
ICAM1
Liquid
ICAM1
Burkitt′s lymphoma cell line Raji.
Protein A
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunoprecipitation
MEM-112
PBS with 15mM sodium azide; pH 7.4
P05362
BB2, P3.58, ICAM-1, cell surface glycoprotein P3.58, intercellular adhesion molecule 1, human rhinovirus receptor, major group rhinovirus receptor
Mouse
IgG1
100 μg
4° C, do not freeze
Primary
3383