Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-1 Mouse anti-Human, FITC, Clone: 1H4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA119616

Catalog No. 01-676-343


Add to cart

Description

Description

CD54 (ICAM-1) is a 90 kD member of the C2 subset of immunoglobulin superfamily. It is a transmembrane molecule with 7 potential N-glycosylated sites, expressed on resting monocytes and endothelial cells and can be upregulated on many other cells, e.g. with lymphokines, on B- and T-lymphocytes, thymocytes, dendritic cells and also on keratinocytes, chondrocytes, as well as epithelial cells. CD54 mediates cell adhesion by binding to integrins CD11a/CD18 (LFA-1) and to CD11b/CD18 (Mac-1). The interaction of CD54 with LFA-1 enhances antigen-specific T-cell activation.
Specifications

Specifications

ICAM-1
Monoclonal
FITC
ICAM1
Liquid
ICAM1
Raji cells and spleen cells fused with NS1 cells.
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
1H4
PBS with 0.2% BSA and 15mM sodium azide
P05362
BB2, P3.58, ICAM-1, cell surface glycoprotein P3.58, intercellular adhesion molecule 1, human rhinovirus receptor, major group rhinovirus receptor
Mouse
IgG2b
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3383
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit