Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD55 Mouse anti-Human, BB515, Clone: IA10, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 565027

Catalog No. BDB565027


Add to cart

Description

Description

The IA10 monoclonal antibody specifically binds to CD55, which is also known as complement decay-accelerating factor (DAF). CD55 is a glycophosphatidylinositol (GPI)-anchored, single chain membrane glycoprotein of approximately 70 kDa that belongs to the regulators of complement activation (RCA) gene family which includes CD21, CD35, and CD46. CD55 is widely expressed on hematopoietic cells including platelets and erythrocytes, as well as on many non-hematopoietic cells, such as, endothelial cells and epithelial cells. CD55 is involved in protecting cells from damage by autologous activated complement complexes. CD55 prevents the amplification steps of the complement cascade by interfering with the assembly of the C3-convertases, C4b2a and C3bBb, and the C5-convertases, C4b2a3b and C3bBb3b.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

Specifications

Specifications

CD55
IA10
DAF; CR; CROM; TC; Complement decay-accelerating factor
Affinity purified
IgG2a, κ
RUO
Primary
Human
Flow Cytometry
BB515
Aqueous buffered solution containing ≤0.09% sodium azide
Murine
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit