Learn More
CD56 Mouse anti-Human, Biotin, Clone: C5.9, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112145
Catalog No. 01-670-864
Add to cart
Description
Description
This antibody recognizes CD56-positive cells. CD56 is a type 1 transmembrane glycoprotein also known as NCAM (Neural Cell Adhesion Molecule). NCAM is expressed on NK cells and a subpopulation of T-cells known as large granular lymphocytes. NCAM appears to play a role in homotypic adhesion of neuronal cells, but the significance of NCAM has not been clearly defined.
Specifications
Specifications
CD56
Monoclonal
Biotin
NCAM1
Liquid
NCAM1
human cell line KG-1a
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
C5.9
PBS with 0.2% carrier protein and 0.08% sodium azide
P13591
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Mouse
IgG2b
100 Tests
4° C, do not freeze
Primary
4684