Learn More
CD61 Mouse anti-Human, Brilliant Violet 650, Clone: VI-PL2, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564172
Catalog No. BDB564172
Add to cart
Description
Description
The VI-PL2 monoclonal antibody specifically binds to CD61. CD61 is a 105 kDa transmembrane glycoprotein that is also known as integrin β3 and platelet glycoprotein IIIa (GPIIIa or GP3A). It is expressed on platelets, megakaryocytes, osteoclasts and endothelia. Integrin β3 associates with gpIIa (CD41) to form the CD41/CD61 complex which mediates platelet adhesion and aggregation. CD61 also associates with CD51 to form the CD51/CD61 complex (vitronectin receptor). CD61 appears to bind to fibrinogen, fibronectin, vWF, vitronectin, and thrombospondin to mediate cell adhesion.
Specifications
Specifications
CD61
VI-PL2
Integrin β3; Integrin beta-3; GP3A; GPIIIa; ITGB3; ITB3
Affinity Purified
IgG1, κ
RUO
Primary
Human
Flow Cytometry
Brilliant Violet 650
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal