Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD61 Mouse anti-Human, Unlabeled, Clone: 1, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611141

 View more versions of this product

Catalog No. BDB611141


Add to cart

Description

Description

Integrins are heterodimeric transmembrane receptors that mediate cell-cell or cell-matrix adhesion. They contain noncovalently associated α and β subunits that consist of a large extracellular region (the ligand-binding domain), a short transmembrane region, and a cytoplasmic domain of varying length. In mammals, at least 17 α subunits and 8 β subunits have been identified and these proteins can heterodimerize to form at least 22 different receptors. Although there is a high degree of redunancy, each integrin has a specific biological function. For example, the β3 subunit associates with αIIb in platelets where this glycoprotein complex acts as a fibrinogen receptor and mediates platelet aggregation. In endothelial cells (EC), β3 complexes with the αv subunit to form the vitronectin receptor. This receptor mediates endothelial cell adhesion to vitronectin, fibrinogen, von Willebrand factor, thrombospondin, laminin, and fibronectin. In confluent EC cultures, the αvβ3 integrin localizes to focal adhesions at the cell body and cell-cell borders. Thus, continued study of individual integrin subunits will provide insights to mechanisms of cell adhesion and signaling.

Host Species: Mouse
Clone: 1
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Integrin β3 aa. 592-712
Formula Weight [Chemical]: 104kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

CD61
1
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Integrin β3 chain
Affinity Purified
Human Integrin β3 aa. 592-712
150μg
Cell Biology
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.