Learn More
anti-CD62L, BV650; Clone: MEL-14; BD
Manufacturer:  BD Biosciences 564108
Catalog No. BDB564108
Add to cart
Description
Description
Applications:
Flow cytometry
Specifications
Specifications
CD62L
MEL-14
Brilliant Violet 650
Affinity purified
C3H/eb mouse B lymphoma 38C-13
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Rat
IgG2a, kappa
Store undiluted at 4deg.C and protected from prolonged exposure to light. Do not freeze
Monoclonal