Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62L Mouse anti-Human, FITC, Clone: 1H3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112151

Catalog No. 01-670-870


Add to cart

Description

Description

The human CD62L is a 74-95 kD glycoprotein member of the selectin family of adhesion receptors. L-Selectin is comprised of an aminoterminal C-type lectin binding domain, an epidermal growth factor-like domain, two short consensus repeat (SCR) sequences homologous to those found in complement binding proteins, a short spacer region, a transmembrane region and a short cytoplasmic region. Human CD62L (L-Selectin) is constitutively expressed on all classes of leukocytes including lymphocytes (except a substantial population of memory T cells), monocytes and polymorphonuclear cells.
Specifications

Specifications

CD62L
Monoclonal
FITC
SELL
Liquid
SELL
human peripheral blood T lymphocytes
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
1H3
PBS with 0.2% carrier protein and 0.08% sodium azide
P14151
SELL, LAM-1, LAM1, LECAM1, LNHR, LSEL, LYAM1, Leu-8, Lyam-1, PLNHR, TQ1, hLHRc
Mouse
IgG2a
100 Tests
4° C, do not freeze
Primary
6402
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit