Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62P Mouse anti-Human, Brilliant Violet 786, Clone: AK-4, BD

Mouse Monoclonal Antibody

$195.00 - $390.00

Specifications

Antigen CD62P
Isotype IgG1, κ
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Conjugate Brilliant Violet 786
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB563715
View Documents
BD Biosciences
563715
100 Tests Each for $390.00
Add to cart
 
BDB563714
View Documents
BD Biosciences
563714
25 Tests Each for $195.00
Add to cart
 
Description

Description

The AK-4 monoclonal antibody specifically binds to CD62P. CD62P is a 140 kDa type I transmembrane glycoprotein that is also known as P-Selectin, Platelet activation-dependent granule membrane protein (PADGEM), or GMP-140. P-Selectin is stored in the α-granules of platelets and the Weibel-Palade bodies of endothelial cells, and is rapidly transported to the plasma membrane upon activation. P-Selectin is thought to mediate the initial adhesive interactions of neutrophils and monocytes with endothelium in inflammatory responses, and of activated platelets to neutrophils and monocytes in hemostasis.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon™ BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).

Specifications

Specifications

CD62P
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Murine
Affinity Purified
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
IgG1, κ
Brilliant Violet 786
Flow Cytometry
RUO
AK-4
P-Selectin; GMP-140; PADGEM

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit