Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD63 Mouse anti-Human, Clone: CLB-180, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA135272

Catalog No. 01-671-235

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.

Specifications

Specifications

CD63
Monoclonal
Unconjugated
CD63
Liquid
CD63
Human cytochrome-B enriched cells.
1 mL
4° C, store in dark
Primary
967
Flow Cytometry, Immunofluorescence
CLB-180
tissue culture supernatant diluted in TBS with 10MG BSA, 10MG BSA and 0.001% thimerosal; pH 8.0
P08962
LAMP-3, ME491, MLA1, OMA81H, TSPAN30
Mouse
IgG1
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit