Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM6 Mouse anti-Human, Clone: 9A6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA117765

Catalog No. 01-670-178


Add to cart

Description

Description

This antibody recognizes native human CEACAM6 protein expressed in transfected mammalian cells.

CEA-related cell adhesion molecule 6 (CEACAM6, NCA) belongs to the carcinoembryonic antigen (CEA) gene family. It encodes a glycosyl phosphatidyl inositol (GPI)-linked glycoprotein with a MW of 90kDa which is strongly expressed on epithelial cells of the fetal and adult gastrointestinal tract, epithelia of glandular tissues, squamous epithelial cell of the tongue, esophagus and cervix as well as on granulocytes. CEACAM6 expression is upregulated in many adenocarcinomas and leukemias. Like all members of the CEA family, it consists of a single N domain, with structural homology to the immunoglobulin variable domains, followed by one immunoglobulin constant-like A and B domain.
Specifications

Specifications

CEACAM6
Monoclonal
1.6 mg/mL
PBS with no preservative; pH 7.2
P40199
CD66c, CEAL, NCA, carcinoembryonic antigen-related cell adhesion molecule 6, non-specific crossreacting antigen, normal cross-reacting antigen
Mouse
IgG1
100 μg
Store at 4°C short term. For long term storage, Store at -20°C, avoiding freeze/thaw cycles.
Primary
4680
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
9A6
Unconjugated
CEACAM6
Liquid
CEACAM6
Tumor cell lines expressing CEACAM6.
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit