Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSG1 Mouse anti-Human, Clone: BAP3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA117763

Catalog No. 01-670-176

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.This product is either new or inactive, and cannot be ordered online.


Description

Description

This antibody recognizes native human PSG protein expressed in transfected mammalian cells.

Human pregcy-specific glycoproteins (PSG), are a group of closely related secreted glycoproteins which are highly expressed in placental syncytiotrophoblast cells of fetal origin. PSG are commonly expressed in tumors of trophoblast origin (hydatidiform mole, choriocarcinoma). They represent the most abundant fetal protein in the maternal blood at term. The eleven human PSG genes (PSG1-PSG11) form together with the carcinoembryonic antigen (CEA)-related cell adhesion molecule (CEACAM) genes the CEACAM gene family. PSG stimulates secretion of TH2-type cytokines from monocytes. CD9 was shown to represent the monocyte receptor for murine PSG17. PSG are thought to modulate the maternal immune system during pregcy thus protecting the semiallotypic fetus from rejection.
Specifications

Specifications

PSG1
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.2
P11464
PSG1, B1G1, CD66f, DHFRP2, FLJ90598, FLJ90654, PBG1, PSBG1, PSG IIA-d, PSGGA, PSGIIA-a, PSGIIA-b, PSGIIA-c
Mouse
IgG1
100 μg
Store at 4°C short term. For long term storage, Store at -20°C, avoiding freeze/thaw cycles.
Primary
5669
Flow Cytometry
BAP3
Unconjugated
PSG1
Liquid
PSG1
Purified human PSG isolated from retroplacental blood.
Protein G
RUO
Antibody
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit