Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD69 Mouse anti-Human, PE, Clone: SMM, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112155

Catalog No. 01-670-874


Add to cart

Description

Description

CD69 (AIM, Active Inducer Molecule) is a gp28/34 disulfide bonded homodimer with a molecular weight of 60 kDa under non-reducing conditions. CD69 contains one or two N linked oligosacaride and the molecule is present on activated platelets. In normal peripheral blood a variable percentage of cells express the CD69 antigen, and it is involved in lymphocyte signal transduction. Expression CD69 is induced upon activation of T lymphocytes, and may play a role in proliferation. Furthermore, the protein may act to transmit signals in natural killer cells and platelets. Alternative splicing results in multiple transcript variants of CD69. Diseases associated with CD69 dysfunction include coccidiodomycosis and asthma.
Specifications

Specifications

CD69
Monoclonal
PE
CD69
Liquid
CD69
White blood cells.
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
SMM
PBS with 0.2% carrier protein and 0.08% sodium azide
Q07108
AIM, BL-AC/P26, CLEC2C, EA1, GP32/28, MLR-3, C-type lectin domain family 2, member C, CD69 antigen
Mouse
IgG1
100 Tests
4° C, do not freeze
Primary
969
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit