Learn More
CD7/CD33 Mouse anti-Human, FITC, PE, Invitrogen™
Antibody Cocktail
Manufacturer:  Invitrogen MA112285
Catalog No. 01-670-980
Add to cart
Description
Description
This antibody is a monoclonal mixture of FITC-conjugated CD7 (clone MG34, isotype IgG2a) and PE-conjugated CD33 (clone FOS, isotype IgG1).
CD7 is a 40kD transmembrane, single-chain glycoprotein. CD7 is expressed by majority of peripheral blood T cells, NK cells and all Thymocytes. The presence or absence of CD7 is a useful marker for the classification of T- and NK-cell maligcies. The CD7 molecule is absent in a subset of human T cells under certain physiologic conditions. Thus an increase in CD7 negative T cells can be seen in some inflammatory skin disease. CD33 is found on granulocyte and macrophage precursors in the bone marrow, but is not on pluripotent stem cells. The protein is also expressed on, and is a useful marker for, peripheral monocytes. It is also useful for distinguishing myelogenous leukaemia cells from lymphoid or erythroid leukaemias.
Specifications
Specifications
CD7/CD33
Cocktail
FITC, PE
CD33
Liquid
CD33, CD7
T-acute lymphoblastic leukemia (T-ALL) cells and FMY9S5 cells containing the CD33 gene
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
FOS
PBS with 0.2% carrier protein and 0.08% sodium azide
P09564, P20138
GP40, LEU-9, TP41, Tp40, FLJ00391, SIGLEC-3, p67
Mouse
IgG1/IgG2a
100 Tests
4° C, do not freeze
Primary
924, 945