Learn More
CD7 Mouse anti-Human, PE, Clone: MG34, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112026
Catalog No. 01-670-749
Add to cart
Description
Description
CD7 (gp40, Leu9) is a 40 kDa member of the immunoglobulin gene superfamily. CD7 contains N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxy-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. Further, CD7 is also found on natural killer cells, a small subpopulation of normal B cells and on maligt B cells. Cross-linking surface CD7 positively modulates T cell and NK cell activity as measured by calcium fluxes, expression of adhesion molecules, cytokine secretion and proliferation. CD7 associates directly with phosphoinositol 3′-kinase. CD7 ligation induces production of D-3 phosphoinositides and tyrosine phosphorylation. Expression of CD7 is an important marker used in leukemia diagnostics.
Specifications
Specifications
CD7
Monoclonal
PE
CD7
Liquid
CD7
T-acute lymphoblastic leukemia (T-ALL) cells
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MG34
PBS with 0.2% gelatin and 0.08% sodium azide
P09564
GP40, LEU-9, TP41, Tp40
Mouse
IgG2a
100 Tests
4° C, do not freeze
Primary
924