Learn More
CD71 Mouse anti-Human, Brilliant Violet 711, Clone: M-A712, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 563767
Description
The M-A712 monoclonal antibody specifically binds to CD71 which is also known as the transferrin receptor (TFR). This type II transmembrane glycoprotein is expressed on cells as a disulfide-linked homodimer comprised of 95 kDa monomers. CD71 is expressed on activated lymphocytes, monocytes, macrophages, erythroid progenitors, brain endothelium, and most proliferating cells. CD71 is not expressed on resting lymphocytes and is upregulated during lymphocyte responses to antigens or mitogens. Through an endocytic pathway, the transferrin receptor mediates cellular iron uptake by binding and internalizing iron that is bound to transferrin. After releasing iron within the low pH endosomal environment, transferrin and its receptor can be recycled to the cell surface.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon™ BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy™5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Specifications
CD71 | |
Monoclonal | |
Brilliant Violet 711 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG2a κ |
Flow Cytometry | |
M-A712 | |
TFR; TFRC; Trfr; TfR1; Transferrin receptor protein 1 | |
T-CLL and Jurkat Cells | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.