Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD71 Rat anti-Mouse, Alexa Fluor 647, Clone: C2, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 563504

Catalog No. BDB563504


Add to cart

Description

Description

The C2 monoclonal antibody specifically binds to CD71, the transferrin receptor. CD71 is a disulfide-linked homodimer of 95-kDa subunits. CD71 mediates one of the cellular mechanisms for iron uptake, and its expression is regulated according to the cell’s iron requirements. It is expressed at high levels on developing erythroid cells, and it is upregulated after mitogenic activation of B or T lymphocytes. The C2 monoclonal antibody selectivity inhibits some types of T- and B-cell activation by down-regulation of transferrin receptor expression, but it does not block binding of transferrin.

Specifications

Specifications

CD71
C2
Alexa Fluor 647
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG1, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Transferrin Receptor; TR; TfR; TfR1; Tfrc; Trfr; Mtvr-1
Affinity Purified
Mouse cell line
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit