Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST6GAL1 Mouse anti-Human, Clone: LN-1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA135289

Catalog No. 01-671-251


Add to cart

Description

Description

A suggested positive control for this product is human tonsil.

Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.
Specifications

Specifications

ST6GAL1
Monoclonal
0.1 mg/mL
PBS with 0.05% sodium azide
P15907
Sialyltransferase 1, SIAT1, ST6GalI, ST6N, B-cell antigen CD75, CMP-N-acetylneuraminate beta-galactosamide alpha-2, 6-sialyltransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2, 6-sialyltransferase 1, ST6Gal I, alpha 2, 6-ST 1, beta-galactoside alpha-2, 6-sialyltransferase 1, sialyltransferase 1
Mouse
IgM
1 mL
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary
6480
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
LN-1
Unconjugated
ST6GAL1
Liquid
ST6GAL1
Lymphocytes/SU-DHL4 cell line.
Purified
RUO
Antibody
Monoclonal
Human
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit