Learn More
anti-CD8, Alexa Fluor 594, Clone: 53-6.7, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB116T025
 View more versions of this product
Catalog No. FAB116T025
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD8
53-6.7
Flow Cytometry 5 uL/10^6 cells
Supernatant
CD8A
Mouse thymus or spleen
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
925
Mouse
Flow Cytometry
Alexa Fluor 594
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD8, CD8A, CD8a molecule, LEU2, p32
Rat
IgG2a
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects both the alpha and alpha ' chains of mouse CD8 (1).