Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CD8, Alexa Fluor 700, Clone: JG12, R&D Systems™
Supplier: R&D Systems FAB7080N025
This item is not returnable.
View return policy
Specifications
CD8 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD8, CD8A, CD8a molecule, LEU2, p32 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
925 | |
Cotton Rat | |
Purified |
Flow Cytometry | |
JG12 | |
Flow Cytometry 5 uL/10^6 cells | |
AAL55392 | |
CD8A | |
NIH-3T3 mouse embryonic fibroblast cell line transfected with cotton rat CD8 alpha and SP2/O mouse melanoma cell line transfected with cotton rat CD8 alpha ., Accession # AAL55392 | |
25 Tests | |
Primary | |
Detects cotton rat CD8 alpha in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title